Learn More
Medchemexpress LLC HY-15910 500mg , 5-BrdU BUdR;BRDU CAS:59-14-3 Purity:98%
SDP
Catalog No. 501492421
Encompass_Preferred
Medchemexpress, HY-15910 500mg 5-BrdU BUdR;BRDU CAS:59-14-3 Formula:C9H11BrN2O5 Purity:98% 5-BrdU is a nucleoside analog, and competes with thymidine for incorporation into DNA, and is used in the detection of proliferating cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2421 Supplier Medchemexpress LLC Supplier No. HY15910500MG
Medchemexpress, HY-15910 500mg 5-BrdU BUdR;BRDU CAS:59-14-3 Formula:C9H11BrN2O5 Purity:98% 5-BrdU is a nucleoside analog, and competes with thymidine for incorporation into DNA, and is used in the detection of proliferating cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.