Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1,2,3-trifluorobenzene, 98%

Catalog No. AAB2060706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2060706 5 g
AAB2060703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2060706 Supplier Thermo Scientific Chemicals Supplier No. B2060706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 138526-69-9 | C6H2BrF3 | 210.981 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138526-69-9
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00070738
InChI Key HKJCELUUIFFSIN-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trifluorobenzene, 3,4,5-trifluorobromobenzene, 1,2,3-trifluoro-5-bromobenzene, 5-bromo-1,2,3-trifluoro-benzene, benzene, 5-bromo-1,2,3-trifluoro, pubchem2304, acmc-1c02k, ksc174s3f, 3,4,5-trifluoro-bromobenzene, 3,4,5-trifluorophenyl bromide
PubChem CID 611409
IUPAC Name 5-bromo-1,2,3-trifluorobenzene
SMILES C1=C(C=C(C(=C1F)F)F)Br

Specifications

Melting Point <-20°C
Density 1.767
Boiling Point 140°C
Flash Point 45°C (113°F)
Refractive Index 1.482
Quantity 5 g
UN Number UN1993
Beilstein 7249191
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 5-Bromo-1,2,3-trifluorobenzene
Show More Show Less
Hazard Category H226-H315-H318-H351
Hazard Statement GHS H Statement
H318-H226-H351-H315
Causes serious eye damage.
Flammable liquid and vapour.
Suspected of causing cancer.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P264b-P281-P303+P361+P353-P305+P351+P338-P308+P313-P310-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 418-480-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.