Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-bromo-1,2-benzisoxazol-3-ylamine, 97%, Thermo Scientific™

Catalog No. MO08226DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO08226DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO08226DA Supplier Thermo Scientific Chemicals Supplier No. MO08226DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 455280-00-9
Molecular Formula C7H5BrN2O
Molecular Weight (g/mol) 213.03
MDL Number MFCD09834823
InChI Key BGMYWBQEKIMHGU-UHFFFAOYSA-N
Synonym 5-bromobenzo d isoxazol-3-ylamine, 5-bromobenzo d isoxazol-3-amine, 5-bromo-1,2-benzisoxazol-3-ylamine, 1,2-benzisoxazol-3-amine, 5-bromo, 3-amino-5-bromobenzo d isoxazole, 5-bromobenzo d isoxazole-3-ylamine, 5-bromanyl-1,2-benzoxazol-3-amine, 1,2-benzisoxazol-3-amine,5-bromo
PubChem CID 33589610
IUPAC Name 5-bromo-1,2-benzoxazol-3-amine
SMILES NC1=NOC2=CC=C(Br)C=C12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1, 2-benzisoxazol-3-ylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.