Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-methyl-1H-indole, 97%, Thermo Scientific™

Catalog No. CC41410DA
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC41410DA 1 g Amber Glass Bottle
CC41410DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC41410DA Supplier Thermo Scientific Chemicals Supplier No. CC41410DA
Add to Cart

Chemical Identifiers

CAS 10075-52-2
Molecular Formula C9H8BrN
Molecular Weight (g/mol) 210.07
MDL Number MFCD00159857
InChI Key SBOITLSQLQGSLO-UHFFFAOYSA-N
PubChem CID 285757
IUPAC Name 5-bromo-1-methylindole
SMILES CN1C=CC2=CC(Br)=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-bromo-1-methyl-1h-indole, 5-bromo-1-methyl-indole, 1h-indole, 5-bromo-1-methyl, 1-methyl-5-bromoindole, 1h-indole,5-bromo-1-methyl, pubchem7946, zlchem 1345, n-methyl-5-bromoindole, 5-bromo-n-methylindole, acmc-2097se
Formula Weight 210.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methyl-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.