Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-1-methylpyridin-2(1H)-one 98.0+%, TCI America™
SDP

Catalog No. B54821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54821G Supplier TCI America Supplier No. B54821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81971-39-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD09972188
InChI Key HHVFVHIUOMMWRN-UHFFFAOYSA-N
PubChem CID 13024027
IUPAC Name 5-bromo-1-methylpyridin-2-one
SMILES CN1C=C(C=CC1=O)Br

Specifications

Melting Point 65°C
Boiling Point 157°C
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-1-methylpyridin-2(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.