Learn More
AdipoGen 5-Bromo-1-pentene
SDP
Supplier:  AdipoGen CDXB0624G025
Encompass_Preferred
Chemical. CAS 1119-51-3. Formula C5H9Br. MW 149.03. 5-Bromo-1-pentene is an organic compound used as building block, starting material or intermediate in organic chemistry. 5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-3-carboxypropyl estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids and is widely used in the fields of medicine, pesticide and material chemistry.
Catalog No. 50-256-1579