Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-1-pentene

Catalog No. 25390125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-390-125G 25 g
25-390-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-390-125G Supplier Sigma Aldrich Supplier No. 25390125G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-bromo-1-pentene was used in stereoselective synthesis of 7α-(3-carboxypropyl) estradiol
  • It was used in preparation of thioacetate 11 of sialic acid having thioglycosidic linkage
  • It was also used as staring material in recent syntheses of dl-histrionicotoxin and benzophenone-containing fatty acids

Specifications

CAS 1119-51-3
Density 1.258 g/mL (at 25°C (literature))
Boiling Point 126°C to 127°C (765 mmHg)
Molecular Formula C5H9Br
Refractive Index n20/D 1.463 (literature)
Linear Formula Br(CH2)3 CH=CH2
MDL Number MFCD00000264
Quantity 25 g
Molecular Weight (g/mol) 149.03
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.