Learn More
Sigma Aldrich 5-Bromo-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 3-chloro-4-fluoroiodobenzene, also known as 2-chloro-1-fluoro-4-iodobenzene [iupac name], is a polyhalobenzene
- It participates in the synthesis of 3-carboxythiophene analogs.3-chloro-4-fluoroiodobenzene may be used in the preparation of 2-chloro-1-(3,5-dimethoxyphenoxy)-4-iodobenzene by reacting with 3,5-dimethoxyphenol in the presence of cesium carbonate
Specifications
Specifications
| Molecular Formula | C14H9BrN2O4S |
| Linear Formula | C14H9BrN2O4S |
| MDL Number | MFCD17171347 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 381.2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.