Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine

Catalog No. ADE0009031G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0009031G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0009031G Supplier Sigma Aldrich Supplier No. ADE0009031G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Menthone is a cyclic oxygenated terpene and its antioxidant potential has been evaluated. it is one of the key active component of danshu capsule (dsc), a medicinal compound in traditional chinese medicine.menthone has been used in the following studies:
  • To prepare the model flavor mix in a study to investigate the release of various aroma compounds in xanthan-thickened food model systems having different viscosities
  • As standard in the quantification of volatile constituents and odour-activity value (oav) in αmarionα and αblack diamondα
  • Modified semisolid agar antifungal susceptibility method (saas) to investigate the anti-fungal activities of various cyclic terpenes against fusarium verticillioides mrc 826

Specifications

CAS 858116-66-2
Molecular Formula C16H25BrN2Si
Linear Formula C16H25BrN2Si
MDL Number MFCD08688597
Quantity 100 mL
Molecular Weight (g/mol) 353.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.