Learn More
Medchemexpress LLC 5-Bromo-1H-indole-2-carboxylic acid | 7254-19-5 | 240.06 | 5 G
SDP
Catalog No. 5000447554
Encompass_Preferred
5-Bromo-1H-indole-2-carboxylic acid is an indole-derived organic compound substituted with bromine and carboxyl groups. It serves as an important intermediate for the synthesis of other active compounds.
  • The dimeric form of this compound exhibits antibacterial activity against Gram-positive bacteria such as *L. monocytogenes*.
Catalog No. 50-004-47554 Supplier Medchemexpress LLC Supplier No. HYW0075935G
5-Bromo-1H-indole-2-carboxylic acid is an indole-derived organic compound substituted with bromine and carboxyl groups. It serves as an important intermediate for the synthesis of other active compounds.
  • The dimeric form of this compound exhibits antibacterial activity against Gram-positive bacteria such as *L. monocytogenes*.