Learn More
Medchemexpress LLC 5-Bromo-1H-pyrazolo[3,4-b]pyridine | 875781-17-2 | 99.8% | C6H4BrN3 | 5 G
SDP
Catalog No. 5000433950
Encompass_Preferred
5-Bromo-1H-pyrazolo[3,4-b]pyridine is a drug intermediate for the synthesis of sulfonamide derivatives. It appears as a solid, white to off-white in color, with a molecular weight of 198.02. This product is intended for research use only.
  • Categorized as a drug intermediate
  • Utilized in the synthesis of sulfonamide derivatives
  • For research use only
  • Solid, white to off-white appearance
  • Molecular weight of 198.02
  • Powder form stable at -20°C for 3 years or 4°C for 2 years
  • When in solvent, stable at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-33950 Supplier Medchemexpress LLC Supplier No. HY690775G
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5-Bromo-1H-pyrazolo[3,4-b]pyridine is a drug intermediate for the synthesis of sulfonamide derivatives. It appears as a solid, white to off-white in color, with a molecular weight of 198.02. This product is intended for research use only.
  • Categorized as a drug intermediate
  • Utilized in the synthesis of sulfonamide derivatives
  • For research use only
  • Solid, white to off-white appearance
  • Molecular weight of 198.02
  • Powder form stable at -20°C for 3 years or 4°C for 2 years
  • When in solvent, stable at -80°C for 6 months or -20°C for 1 month