Learn More
Medchemexpress LLC 5-bromo-2-(2,6-dioxopiperidin-3-yl)-4-fluoroisoindoline-1,3-dione | 2740657-28-5 | MFCD34180776 | >98.0% | 355.12 g/mol | C13H8BrFN2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW453715100MG
Encompass_Preferred
Thalidomide-4-F,5-Br is a thalidomide analogue used as a cereblon ligand in chemical biology and targeted protein degradation research. The compound is supplied as a solid research reagent (CAS 2740657-28-5) with a reported formula of C13H8BrFN2O4 and reported purity >98% in supplier documentation. Typical applications include PROTAC development, biochemical assays, and studies of E3 ligase-mediated ubiquitination.
  • Acts as a cereblon ligand for recruiting E3 ubiquitin ligase.
  • Useful in PROTAC design and targeted protein degradation studies.
  • High reported purity suitable for research applications.
  • Solid form stable for standard laboratory storage conditions.
  • CAS-registered for unambiguous chemical identification.
Catalog No. 50-002-81499