Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-bromo-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione | 26166-92-7 | MFCD30188073 | 98.4% | 337.13 | C13H9BrN2O4 | 100 MG
SDP

Catalog No. 5000210245
Encompass_Preferred

Thalidomide-5-Br is a brominated thalidomide derivative supplied for research use in studies of inflammation, tumor biology, and neurology. It is provided as a solid research reagent; consult lot-specific documentation for analytical details.

  • Contains a bromodomain; used as a tool compound in biochemical and pharmacological research.
  • Reported purity approximately 98% (see product documentation for lot-specific certificate of analysis).
  • Solid, off-white to gray appearance suitable for analytical and synthetic applications.
  • Storage recommendations: room temperature for shipping; in solution store at -80°C (2 years) or -20°C (1 year).
  • Available in multiple pack sizes including 100 mg.

Catalog No. 50-002-10245 Supplier Medchemexpress LLC Supplier No. HYW069604100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-5-Br is a brominated thalidomide derivative supplied for research use in studies of inflammation, tumor biology, and neurology. It is provided as a solid research reagent; consult lot-specific documentation for analytical details.

  • Contains a bromodomain; used as a tool compound in biochemical and pharmacological research.
  • Reported purity approximately 98% (see product documentation for lot-specific certificate of analysis).
  • Solid, off-white to gray appearance suitable for analytical and synthetic applications.
  • Storage recommendations: room temperature for shipping; in solution store at -80°C (2 years) or -20°C (1 year).
  • Available in multiple pack sizes including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.