Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-(2-thienyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B4885200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4885200MG 200 mg
B48851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4885200MG Supplier TCI America Supplier No. B4885200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91891-74-6
Molecular Formula C9H6BrNS
Molecular Weight (g/mol) 240.12
MDL Number MFCD09037419
InChI Key POYKHSYCYNRTKC-UHFFFAOYSA-N
Synonym 5-bromo-2-2-thienyl pyridine, 5-bromo-2-thiophen-2-yl pyridine, 5-bromo-2-thien-2-yl pyridine, 2-2-thienyl-5-bromopyridine, 2-5-bromo-2-pyridyl thiophene
PubChem CID 13238694
IUPAC Name 5-bromo-2-(thiophen-2-yl)pyridine
SMILES BrC1=CN=C(C=C1)C1=CC=CS1

Specifications

Melting Point 81°C
Quantity 200 mg
Formula Weight 240.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-(2-thienyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.