Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,3-dihydrobenzofuran 98.0+%, TCI America™
SDP

Catalog No. B43185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B43185G Supplier TCI America Supplier No. B43185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66826-78-6
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD02677716
InChI Key UDWFSJAYXTXMLM-UHFFFAOYSA-N
Synonym 5-bromo-2,3-dihydrobenzofuran, 5-bromo-2,3-dihydrobenzo b furan, 5-bromocoumaran, benzofuran, 5-bromo-2,3-dihydro, 2,3-dihydro-5-bromobenzofuran, buttpark 36\18-16, pubchem7055, 5-bromo-2,3 dihydrobenzofuran, acmc-1b981, 5-bromo-2,3-dihydro-benzofuran
PubChem CID 2776159
IUPAC Name 5-bromo-2,3-dihydro-1-benzofuran
SMILES BrC1=CC2=C(OCC2)C=C1

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2,3-dihydrobenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.