Learn More
Medchemexpress LLC 5-bromo-2-(4-fluorophenyl)-3-[4-(methylsulfonyl)phenyl]-thiophene | 88149-94-4 | MFCD02684527 | 99.8% | 411.31 | C17H12BrFO2S2 | 5 MG
SDP
Catalog No. 5000244742
Encompass_Preferred
DuP-697 (CAS 88149-94-4) is a selective, irreversible cyclooxygenase-2 (COX-2) inhibitor provided as a solid research reagent for biochemical and cell-based assays. It exhibits potent inhibition of human COX-2 with marked selectivity over COX-1 and is used to investigate prostaglandin-mediated inflammation and related pharmacology.
  • Selective, irreversible COX-2 inhibitor with hCOX-2 IC50 = 10 nM.
  • High purity solid suitable for biochemical and cell-based assays.
  • Demonstrates activity in cellular models and animal inflammation assays.
  • Stable as a powder at -20°C for long-term storage; limit solvent storage to -80°C for extended periods.
  • Useful tool compound for studies of prostaglandin synthesis and inflammation pathways.
Catalog No. 50-002-44742 Supplier Medchemexpress LLC Supplier No. HY1033875MG
DuP-697 (CAS 88149-94-4) is a selective, irreversible cyclooxygenase-2 (COX-2) inhibitor provided as a solid research reagent for biochemical and cell-based assays. It exhibits potent inhibition of human COX-2 with marked selectivity over COX-1 and is used to investigate prostaglandin-mediated inflammation and related pharmacology.
  • Selective, irreversible COX-2 inhibitor with hCOX-2 IC50 = 10 nM.
  • High purity solid suitable for biochemical and cell-based assays.
  • Demonstrates activity in cellular models and animal inflammation assays.
  • Stable as a powder at -20°C for long-term storage; limit solvent storage to -80°C for extended periods.
  • Useful tool compound for studies of prostaglandin synthesis and inflammation pathways.