Learn More
Medchemexpress LLC 5-bromo-2-(4-fluorophenyl)-3-(4-methylsulfonylphenyl)thiophene | 88149-94-4 | MFCD02684527 | 99.8% | 411.31 g·mol⁻¹ | C17H12BrFO2S2 | 10 MG
SDP
Catalog No. 5000244718
Encompass_Preferred
DuP-697 (CAS 88149-94-4) is a selective, irreversible cyclooxygenase-2 (COX-2) inhibitor used as a research chemical. It inhibits prostaglandin synthesis and has demonstrated antiproliferative, antiangiogenic, and antipyretic effects in cellular studies. The compound is provided as a high-purity solid for biochemical assays and preclinical research; it is not intended for human or veterinary use.
  • Selective, irreversible COX-2 inhibition (IC50 ≈ 10 nM for human COX-2)
  • Minimal COX-1 activity (IC50 ≈ 800 nM for human COX-1)
  • High purity (≈99.8%) suitable for analytical and biological studies
  • Molecular weight 411.31 g·mol⁻¹; formula C17H12BrFO2S2
  • Suitable for biochemical assays and cell-based research; research use only
Catalog No. 50-002-44718 Supplier Medchemexpress LLC Supplier No. HY10338710MG
Add to Cart
Add to Cart
DuP-697 (CAS 88149-94-4) is a selective, irreversible cyclooxygenase-2 (COX-2) inhibitor used as a research chemical. It inhibits prostaglandin synthesis and has demonstrated antiproliferative, antiangiogenic, and antipyretic effects in cellular studies. The compound is provided as a high-purity solid for biochemical assays and preclinical research; it is not intended for human or veterinary use.
  • Selective, irreversible COX-2 inhibition (IC50 ≈ 10 nM for human COX-2)
  • Minimal COX-1 activity (IC50 ≈ 800 nM for human COX-1)
  • High purity (≈99.8%) suitable for analytical and biological studies
  • Molecular weight 411.31 g·mol⁻¹; formula C17H12BrFO2S2
  • Suitable for biochemical assays and cell-based research; research use only