Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-benzoxazolinone 98.0+%, TCI America™
SDP

Catalog No. B44381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44381G 1 g
B44385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44381G Supplier TCI America Supplier No. B44381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14733-73-4
Molecular Formula C7H4BrNO2
Molecular Weight (g/mol) 214.02
MDL Number MFCD01664246
InChI Key DMHTZWJRUUOALC-UHFFFAOYSA-N
Synonym 5-Bromo-2-benzoxazolone, 5-Bromo-2-hydroxybenzoxazole
PubChem CID 26853
IUPAC Name 5-bromo-2,3-dihydro-1,3-benzoxazol-2-one
SMILES BrC1=CC=C2OC(=O)NC2=C1

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 1 g
Formula Weight 214.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-benzoxazolinone
RTECSNumber DM4950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.