Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chloro-3-fluoropyridine 98.0+%, TCI America™
SDP

Catalog No. B50895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50895G 5 g
B508925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50895G Supplier TCI America Supplier No. B50895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 831203-13-5
Molecular Formula C5H4BrFN2
Molecular Weight (g/mol) 191.00
MDL Number MFCD03411574
InChI Key CLYOMCMDBNNGSM-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro-5-bromopyridine, 5-bromo-2-chloro-3-fluoro-pyridine, pyridine,5-bromo-2-chloro-3-fluoro, pyridine, 5-bromo-2-chloro-3-fluoro, pubchem1163, acmc-209pr9, ksc495k9j, 3-bromo-6-chloro-5-fluoropyridine, 3-bromo-6-chloro-5-fluoro-pyridine, 5-bromo-2-chloro-3-fluoro pyridine
PubChem CID 2763285
IUPAC Name 5-bromo-2-fluoropyridin-3-amine
SMILES NC1=CC(Br)=CN=C1F

Specifications

Melting Point 52°C
Color White
Quantity 5 g
Formula Weight 210.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Bromo-2-chloro-3-fluoropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.