Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-chloro-4-(dimethylamino)pyrimidine

Catalog No. SYX002731G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002731G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002731G Supplier Sigma Aldrich Supplier No. SYX002731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tris(2-chloroethyl) phosphate is used as a flame retardant in plastics, especially inflexible foams used in automobiles and furniture and in rigid foams used for building insulation.Tris(2-chloroethyl) phosphate was used in dynamic air sampling of airborne organophosphate triesters using a solid-phase microextraction device.

Specifications

CAS 57054-86-1
Molecular Formula C6H7BrClN3
Linear Formula C6H7BrClN3
MDL Number MFCD07781214
Quantity 25 g
Molecular Weight (g/mol) 236.5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.