Learn More
Medchemexpress LLC 5-bromo-2-chloro-4-methylpyridine | 778611-64-6 | MFCD03427663 | 98.6% | 206.47 | C6H5BrClN | 25 G
SDP
Catalog No. 5000303585
Encompass_Preferred
5-Bromo-2-chloro-4-methylpyridine (CAS 778611-64-6) is an organic small-molecule reagent used as a building block in medicinal chemistry and synthetic organic research. It is supplied for laboratory research and synthesis applications where a halogenated pyridine intermediate is required.
  • High purity suitable for research (98.62%).
  • Well-characterized: molecular formula C6H5BrClN, MW 206.47.
  • Available in convenient small-scale packaging for lab use (10 g, 25 g, 50 g).
  • Useful as a synthetic intermediate in heterocycle and pharmaceutical synthesis.
  • Stable under standard storage conditions for typical laboratory reagents.
Catalog No. 50-003-03585 Supplier Medchemexpress LLC Supplier No. 25GHYW014584
Add to Cart
Add to Cart
5-Bromo-2-chloro-4-methylpyridine (CAS 778611-64-6) is an organic small-molecule reagent used as a building block in medicinal chemistry and synthetic organic research. It is supplied for laboratory research and synthesis applications where a halogenated pyridine intermediate is required.
  • High purity suitable for research (98.62%).
  • Well-characterized: molecular formula C6H5BrClN, MW 206.47.
  • Available in convenient small-scale packaging for lab use (10 g, 25 g, 50 g).
  • Useful as a synthetic intermediate in heterocycle and pharmaceutical synthesis.
  • Stable under standard storage conditions for typical laboratory reagents.