Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-2-chloropyridine | 53939-30-3 | 192.44 | 1 ML
SDP

Catalog No. 5000445705
Encompass_Preferred

5-Bromo-2-chloropyridine is a biochemical reagent used as a biological material or organic compound for life science related research. This product is supplied as a 10 mM solution in 1 mL of DMSO, ready for use.

  • High purity of 99.99%
  • Molecular weight: 192.44
  • Chemical formula: C5H3BrClN
  • Appearance: White to light yellow solid
  • Solubility in DMSO: 100 mg/mL
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage for solution: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-45705 Supplier Medchemexpress LLC Supplier No. HYW00276710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Bromo-2-chloropyridine is a biochemical reagent used as a biological material or organic compound for life science related research. This product is supplied as a 10 mM solution in 1 mL of DMSO, ready for use.

  • High purity of 99.99%
  • Molecular weight: 192.44
  • Chemical formula: C5H3BrClN
  • Appearance: White to light yellow solid
  • Solubility in DMSO: 100 mg/mL
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage for solution: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.