Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-2-chloropyridine | 53939-30-3 | 192.44 | 100 G
SDP

Catalog No. 5000445704
Encompass_Preferred

5-Bromo-2-chloropyridine is a biochemical reagent and organic compound designed for life science research. It serves as a laboratory chemical and is suitable for the manufacture of various substances. This product is for research use only and not intended for medical applications.

  • High purity of 99.99%
  • Molecular formula C₅H₃BrClN
  • Molecular weight of 192.44
  • CAS number 53939-30-3
  • Appearance as a white to light yellow solid
  • 1H NMR spectrum consistent with structure
  • Low water content (0.12% KF)
  • Melting point 65-69 °C
  • Boiling point 208.1 °C

Catalog No. 50-004-45704 Supplier Medchemexpress LLC Supplier No. HYW002767100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-2-chloropyridine is a biochemical reagent and organic compound designed for life science research. It serves as a laboratory chemical and is suitable for the manufacture of various substances. This product is for research use only and not intended for medical applications.

  • High purity of 99.99%
  • Molecular formula C₅H₃BrClN
  • Molecular weight of 192.44
  • CAS number 53939-30-3
  • Appearance as a white to light yellow solid
  • 1H NMR spectrum consistent with structure
  • Low water content (0.12% KF)
  • Melting point 65-69 °C
  • Boiling point 208.1 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.