Learn More
Medchemexpress LLC 5-Bromo-2-chloropyrimidine | 32779-36-5 | 99.9% | C4H2BrClN2 | 100 G
SDP
Catalog No. 5000434490
Encompass_Preferred
5-Bromo-2-chloropyrimidine is a biochemical reagent utilized in life science research as a biological material or organic compound. It has a molecular weight of 193.43 and a high purity of 99.92%. This compound is typically stored in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition. Recommended storage temperature for the solid form is 4°C, sealed, away from moisture and light. If in solution, it should be stored at -80°C for up to 6 months or -20°C for 1 month, also under sealed conditions away from moisture and light.
  • Biochemical reagent for life science research
  • High purity
  • Stable under recommended storage conditions
  • Organic compound
Catalog No. 50-004-34490 Supplier Medchemexpress LLC Supplier No. HY76897100G
5-Bromo-2-chloropyrimidine is a biochemical reagent utilized in life science research as a biological material or organic compound. It has a molecular weight of 193.43 and a high purity of 99.92%. This compound is typically stored in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition. Recommended storage temperature for the solid form is 4°C, sealed, away from moisture and light. If in solution, it should be stored at -80°C for up to 6 months or -20°C for 1 month, also under sealed conditions away from moisture and light.
  • Biochemical reagent for life science research
  • High purity
  • Stable under recommended storage conditions
  • Organic compound