Learn More
Accela Chembio Inc 5-bromo-2-cyanopyridine | 100g | 97483-77-7 | MFCD00234144 | 97% | Shelf Life: 1620 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50479207
Encompass_Preferred
5-bromo-2-cyanopyridine | 100g | 97483-77-7 | MFCD00234144 | 97% | Shelf Life: 1620 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-479-207 Supplier Accela Chembio Inc Supplier No. SY003393100G
5-bromo-2-cyanopyridine | 100g | 97483-77-7 | MFCD00234144 | 97% | Shelf Life: 1620 Days | Light Sensitive/nitrogen Or Argon/+4