Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromo-2'-deoxy-2'-fluorouridine | 55612-18-5 | 97.0% | 325.09 | 1 G
SDP

Catalog No. 5000359428
Encompass_Preferred

5-Bromo-2'-deoxy-2'-fluorouridine is a chemical compound known as a 5-halo analog with potential as an antiviral agent. This product is a solid, appearing white to off-white, and is stable under recommended storage conditions.

  • Designed for laboratory chemical applications
  • Molecular formula: C9H10BrFN2O5
  • Store at 4°C under nitrogen in a cool, well-ventilated area
  • Avoid direct sunlight and sources of ignition
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month, under nitrogen
  • Can be shipped at room temperature if the duration is less than 2 weeks

Catalog No. 50-003-59428 Supplier Medchemexpress LLC Supplier No. HY1489571G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromo-2'-deoxy-2'-fluorouridine is a chemical compound known as a 5-halo analog with potential as an antiviral agent. This product is a solid, appearing white to off-white, and is stable under recommended storage conditions.

  • Designed for laboratory chemical applications
  • Molecular formula: C9H10BrFN2O5
  • Store at 4°C under nitrogen in a cool, well-ventilated area
  • Avoid direct sunlight and sources of ignition
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month, under nitrogen
  • Can be shipped at room temperature if the duration is less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.