Learn More
Medchemexpress LLC 5-Bromo-2'-deoxycytidine | 1022-79-3 | 99.8% | 306.11 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW011376500MG
Encompass_Preferred
5-Bromo-2'-deoxycytidine is a DNA photosensitizer. It modifies double-stranded DNA and helps determine DNA damage using 300 nm photons. This product is for research use only.
  • DNA photosensitizer
  • Modifies double-stranded DNA
  • Determines DNA damage using 300 nm photons
Catalog No. 50-004-53044