Learn More
Cayman Chemical 5-Bromo-2-deoxyurIdIn 1g
SDP
Catalog No. 502968511
Encompass_Preferred
A thymidine nucleoside derivative commonly used to identify proliferating cells; intraperitoneal injection of BrdU can be used in combination with neuronal markers to detect adult neurogenesis in animal models; commonly used as a marker of proliferation of cancer cells in vitro
Catalog No. 50-296-8511 Supplier Cayman Chemical Supplier No. 155801 G
A thymidine nucleoside derivative commonly used to identify proliferating cells; intraperitoneal injection of BrdU can be used in combination with neuronal markers to detect adult neurogenesis in animal models; commonly used as a marker of proliferation of cancer cells in vitro