Learn More
Cayman Chemical 5-Bromo-2-deoxyurIdIn 500mg
SDP
Catalog No. 502970844
Encompass_Preferred
A thymidine nucleoside derivative commonly used to identify proliferating cells; intraperitoneal injection of BrdU can be used in combination with neuronal markers to detect adult neurogenesis in animal models; commonly used as a marker of proliferation of cancer cells in vitro
Catalog No. 50-297-0844 Supplier Cayman Chemical Supplier No. 15580500 MG
A thymidine nucleoside derivative commonly used to identify proliferating cells; intraperitoneal injection of BrdU can be used in combination with neuronal markers to detect adult neurogenesis in animal models; commonly used as a marker of proliferation of cancer cells in vitro