Learn More
Bioworld 5-Bromo-2'-deoxyuridine, 5 g
Catalog No. 50255660
Encompass_Preferred
It is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation. Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase.Appearance: White powder.ChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 191-194 °C (dec.)(lit.)
Catalog No. 50-255-660 Supplier Bioworld Supplier No. 402900003
It is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation. Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase.Appearance: White powder.ChEBI : Not available.Crystal_Structure : Not available.InChI : Not available.InChIKey : Not available.Melting Point: 191-194 °C (dec.)(lit.)