Learn More
Alkali Scientific 5 Bromo 2' deoxyuridine, 250 Milligrams
SDP
Catalog No. 5000659875
Encompass_Preferred
Bromo Deoxyuridine (BrdU) is a nucleoside analog used in DNA labeling, supplied in 250mg quantity. BrdU is commonly incorporated into newly synthesized DNA during cell division, allowing researchers to track DNA replication and study cell proliferation. This compound is used in cellular biology and molecular research, particularly in assays that require the monitoring of cell cycle progression. BrdU labeling is a powerful technique for identifying proliferating cells in tissues, and it is widely used in cancer research, immunology, and neurobiology. It is also used for assessing DNA synthesis rates and conducting flow cytometry experiments related to cell division and DNA repair processes.
Catalog No. 50-006-59875 Supplier Alkali Scientific Supplier No. BRO004CP
Bromo Deoxyuridine (BrdU) is a nucleoside analog used in DNA labeling, supplied in 250mg quantity. BrdU is commonly incorporated into newly synthesized DNA during cell division, allowing researchers to track DNA replication and study cell proliferation. This compound is used in cellular biology and molecular research, particularly in assays that require the monitoring of cell cycle progression. BrdU labeling is a powerful technique for identifying proliferating cells in tissues, and it is widely used in cancer research, immunology, and neurobiology. It is also used for assessing DNA synthesis rates and conducting flow cytometry experiments related to cell division and DNA repair processes.