Learn More
Research Products International Corp 5-Bromo-2-Deoxyuridine 250 MG
SDP
Catalog No. 502230319
Encompass_Preferred
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.
Catalog No. 50-223-0319 Supplier Research Products International Corp Supplier No. B742000.25
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.