Learn More
Sigma Aldrich Fine Chemicals Biosciences 5 Bromo 2 deoxyuridine 5 t5MG
Catalog No. 502903963
Encompass
5-Bromo-2AE-deoxyuridine 5AE-triphosphate is used for incorporation into DNA for studying normal and tumor cell proliferation profiles.
Catalog No. 50-290-3963 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B06315MG
5-Bromo-2AE-deoxyuridine 5AE-triphosphate is used for incorporation into DNA for studying normal and tumor cell proliferation profiles.