Learn More
Research Products International Corp 5-Bromo-2'-Deoxyuridine, 500 Milligrams
SDP
Catalog No. 50213199
Encompass_Preferred
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.
Catalog No. 50-213-199 Supplier Research Products International Corp Supplier No. B742000.5
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.