Learn More
Research Products International Corp 5-Bromo-2'-Deoxyuridine, 5 Grams
SDP
Catalog No. 50213201
Encompass_Preferred
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.
Catalog No. 50-213-201 Supplier Research Products International Corp Supplier No. B742005.0
Thymidine analog used for study of DNA synthesis. This analog selectively incorporates into cellular DNA during S-phase. Supplied as white powder.