Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-bromo-2-ethoxy-4-pyridineboronic acid

Catalog No. CDS006725 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS006725 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS006725 Supplier Sigma Aldrich Supplier No. CDS00672525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl methoxyacetate was used:
  • As acyl donor during the preparation of enantiomers of several phenylethylamines
  • As acylation reagent for the aminolysis of 1-phenylethanamine
  • In an industrial, lipase-catalysed kinetic resolution of primary amine

Specifications

Molecular Formula C7H9BBrNO3
Linear Formula C7H9BBrNO3
MDL Number MFCD07783887
Quantity 10 g
Molecular Weight (g/mol) 245.87
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.