Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-ethoxypyrimidine

Catalog No. SYX002971G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002971G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002971G Supplier Sigma Aldrich Supplier No. SYX002971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3,4-dichlorobenzyl chloride has been used:
  • In friedel-crafts synthesis of 4-(3,4-dichlorophenyl)-3,4-dihydro-1(2h)-naphthalenone, a key intermediate for the synthesis of sertraline
  • As alkylating agent in synthesis of poly(ether ketone)s having pendant sulfonic acid groups
  • In preparation of 5,6-dichloro-2-(3,4-dichlorobenzylthio)benzimidazole

Specifications

CAS 17758-11-1
Molecular Formula C6H7BrN2O
Linear Formula C6H7BrN2O
MDL Number MFCD07375135
Quantity 100 g
Molecular Weight (g/mol) 203.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.