Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-ethyl-3-iodo-1H-pyrrolo[2,3-b]pyridine

Catalog No. ADE0010631G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0010631G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0010631G Supplier Sigma Aldrich Supplier No. ADE0010631G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Chloromethyl-2,2-dimethyl-1,3-dioxolane is a ketal that can be synthesized by the reaction of 3-chloro-propane-1,2-diol (3-MCPD) with acetone in the presence of p-toluene sulfonic acid.4-Chloromethyl-2,2-dimethyl-1,3-dioxolane may be used in the preparation of 2,2-dimethyl-4-methylene-1,3-dioxolane via dehydrochlorination.

Specifications

CAS 1228666-12-3
Molecular Formula C9H8BrIN2
Linear Formula C9H8BrIN2
MDL Number MFCD16628255
Quantity 25 g
Molecular Weight (g/mol) 350.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.