Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluoro-4-methylpyridine, 96%, Thermo Scientific™

Catalog No. AAH6143114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6143114 25 g
AAH6143106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6143114 Supplier Thermo Scientific Chemicals Supplier No. H6143114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-2-fluoro-4-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864830-16-0
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.015
MDL Number MFCD09839213
InChI Key PFODFMZCBQUYNG-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-4-picoline, 5-bromo-2-fluoro-4-methyl-pyridine, 2-fluoro-4-methyl-5-bromopyridine, pubchem3547, acmc-209q9w, ksc495s8j
PubChem CID 23157062
IUPAC Name 5-bromo-2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)F

Specifications

Color Colorless to Yellow
Refractive Index 1.53
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 190.01
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-fluoro-4-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.