Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluoro-m-xylene 97.0+%, TCI America™
SDP

Catalog No. B43705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43705G 5 g
B43701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43705G Supplier TCI America Supplier No. B43705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 99725-44-7 | C8H8BrF | 203.054 g/mol

Chemical Identifiers

CAS 99725-44-7
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD01320701
InChI Key ZXPHUVHMBKRRJF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoro-m-xylene, 5-bromo-2-fluoro-3-xylene, 4-bromo-2,6-dimethylfluorobenzene, 1-bromo-4-fluoro-3,5-dimethylbenzene, 5-bromo-2-fluoro-1,3-xylene, benzene, 5-bromo-2-fluoro-1,3-dimethyl, 3,5-dimethyl-4-fluorobromobenzene, 5-bromo-2-fluoro-1,3-dimethyl-benzene, ksc486i9j
PubChem CID 2736298
IUPAC Name 5-bromo-2-fluoro-1,3-dimethylbenzene
SMILES CC1=CC(=CC(=C1F)C)Br

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 89°C
Formula Weight 203.05
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 5-Bromo-2-fluoro-m-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.