Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluoropyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAL1991506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1991506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1991506 Supplier Thermo Scientific Chemicals Supplier No. L1991506
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501435-91-2
Molecular Formula C5H4BBrFNO2
Molecular Weight (g/mol) 219.80
MDL Number MFCD04039317
InChI Key YAXKWQSQZIVTFC-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoropyridine-3-boronic acid, 5-bromo-2-fluoropyridin-3-yl boronic acid, 5-bromo-2-fluoro-3-pyridylboronic acid, 5-bromo-2-fluoro-3-pyridineboronic acid, 2-fluoro-5-bromopyridine-3-boronic acid, boronic acid, 5-bromo-2-fluoro-3-pyridinyl, 5-bromo-2-fluoro-3-pyridinyl-boronic acid, pubchem5317, acmc-1axfu, ksc495q8d
PubChem CID 2783242
IUPAC Name (5-bromo-2-fluoropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC(Br)=CN=C1F

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 219.81
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluoropyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.