Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-fluoropyridine-3-carboxaldehyde, 95%, Thermo Scientific™

Catalog No. AAH61123MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61123MD 250 mg
AAH6112303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61123MD Supplier Thermo Scientific Chemicals Supplier No. H61123MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875781-15-0
Molecular Formula C6H3BrFNO
Molecular Weight (g/mol) 203.998
MDL Number MFCD09842440
InChI Key MUYVOGAJRCBWCY-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoronicotinaldehyde, 5-bromo-2-fluoropyridine-3-carboxaldehyde, 5-bromo-2-fluoro-3-pyridinecarboxaldehyde, 5-bromo-2-fluoro-pyridine-3-carbaldehyde, 5-bromo-2-fluoro-3-formylpyridine, 3-pyridinecarboxaldehyde, 5-bromo-2-fluoro, pubchem17162, acmc-209qoh, ksc495s8t
PubChem CID 45480192
IUPAC Name 5-bromo-2-fluoropyridine-3-carbaldehyde
SMILES C1=C(C(=NC=C1Br)F)C=O

Specifications

Color White to Yellow
Quantity 250 mg
Beilstein 14515665
Formula Weight 204
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Bromo-2-fluoropyridine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.