Learn More
Medchemexpress LLC 5-Bromo-2-fluoropyridine | 766-11-0 | 175.99 | 25 G
SDP
Catalog No. 5000449458
Encompass_Preferred
5-Bromo-2-fluoropyridine is a biochemical reagent used as a biological material or organic compound for life science research. It readily reacts with amines to synthesize halogenated pyridine and pyrimidine.
  • Suitable for life science-related research
  • Reacts with amines to form halogenated pyridine and pyrimidine
  • Can serve as a model substrate to investigate the efficiency and selectivity of successive SNAr substitution reactions
  • Can serve as a model substrate to investigate transition-metal-catalyzed cross-coupling reactions without needing an intermediate purification step
Catalog No. 50-004-49458 Supplier Medchemexpress LLC Supplier No. 25GHYW008137
Add to Cart
Add to Cart
5-Bromo-2-fluoropyridine is a biochemical reagent used as a biological material or organic compound for life science research. It readily reacts with amines to synthesize halogenated pyridine and pyrimidine.
  • Suitable for life science-related research
  • Reacts with amines to form halogenated pyridine and pyrimidine
  • Can serve as a model substrate to investigate the efficiency and selectivity of successive SNAr substitution reactions
  • Can serve as a model substrate to investigate transition-metal-catalyzed cross-coupling reactions without needing an intermediate purification step