Learn More
Medchemexpress LLC 5-Bromo-2-fluoropyridine | 766-11-0 | 99.2% | 175.99 | 10 G
SDP
Catalog No. 5000404361
Encompass_Preferred
5-Bromo-2-fluoropyridine is a biochemical reagent applicable as a biological material or organic compound in life science research. It readily reacts with amines to form halogenated pyridine and pyrimidine.
  • Serves as a model substrate for investigating efficiency and selectivity in successive SNAr substitution reactions.
  • Used in transition-metal-catalyzed cross-coupling reactions without requiring an intermediate purification step.
  • Applicable as a biological material or organic compound for life science research.
Catalog No. 50-004-04361 Supplier Medchemexpress LLC Supplier No. HY3274610G
Add to Cart
Add to Cart
5-Bromo-2-fluoropyridine is a biochemical reagent applicable as a biological material or organic compound in life science research. It readily reacts with amines to form halogenated pyridine and pyrimidine.
  • Serves as a model substrate for investigating efficiency and selectivity in successive SNAr substitution reactions.
  • Used in transition-metal-catalyzed cross-coupling reactions without requiring an intermediate purification step.
  • Applicable as a biological material or organic compound for life science research.