Learn More
Medchemexpress LLC 5-Bromo-2-fluoropyridine | 766-11-0 | 99.2% | 175.99 | 100 G
SDP
Catalog No. 5000401441
Encompass_Preferred
5-Bromo-2-fluoropyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It easily reacts with amines to synthesize halogenated pyridine and pyrimidine. This compound can also be used as a model substrate to explore efficiency and selectivity through successive SNAr substitution reactions and transition-metal-catalyzed cross-coupling reactions without an intervening purification step.
  • Biochemical reagent for life science research
  • Reacts with amines to synthesize halogenated pyridine and pyrimidine
  • Can serve as a model substrate for SNAr substitution and transition-metal-catalyzed cross-coupling reactions
Catalog No. 50-004-01441 Supplier Medchemexpress LLC Supplier No. HY32746100G
5-Bromo-2-fluoropyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It easily reacts with amines to synthesize halogenated pyridine and pyrimidine. This compound can also be used as a model substrate to explore efficiency and selectivity through successive SNAr substitution reactions and transition-metal-catalyzed cross-coupling reactions without an intervening purification step.
  • Biochemical reagent for life science research
  • Reacts with amines to synthesize halogenated pyridine and pyrimidine
  • Can serve as a model substrate for SNAr substitution and transition-metal-catalyzed cross-coupling reactions