Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromo-2-hydroxybenzenecarbothioamide

Catalog No. OTV0002631G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0002631G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0002631G Supplier Sigma Aldrich Supplier No. OTV0002631G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ammonium thioglycolate solution can be used in the synthesis of:
  • Thiol functionalized eggshell membrane applicable as adsorbent material for the removal of cr(vi), hg(ii), cu(ii), pb(ii), cd(ii), and ag(i) from aqueous water
  • Cellulose-rhodamine b methacrylamide (cellulose-rhbma).it can also be used as a reagent to reduce disulfide bonds in keratin

Specifications

Molecular Formula C7H6BrNOS
Linear Formula C7H6BrNOS
MDL Number MFCD09718404
Quantity 1 L
Molecular Weight (g/mol) 232.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.