Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-hydroxybenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
2316-64-5
C7H7BrO2
203.04
MFCD00004618
KNKRHSVKIORZQB-UHFFFAOYSA-N
5-bromo-2-hydroxybenzyl alcohol,4-bromo-2-hydroxymethyl phenol,bromosaligenin,5-bromosalicyl alcohol,bromsalizol,5-bromosaligenin,5-bromo-2-hydroxybenzylalcohol,2-hydroxymethyl-4-bromophenol,benzenemethanol, 5-bromo-2-hydroxy,unii-y1uz53815e
75342
4-bromo-2-(hydroxymethyl)phenol
C1=CC(=C(C=C1Br)CO)O
2316-64-5
C7H7BrO2
203.04
MFCD00004618
KNKRHSVKIORZQB-UHFFFAOYSA-N
5-bromo-2-hydroxybenzyl alcohol,4-bromo-2-hydroxymethyl phenol,bromosaligenin,5-bromosalicyl alcohol,bromsalizol,5-bromosaligenin,5-bromo-2-hydroxybenzylalcohol,2-hydroxymethyl-4-bromophenol,benzenemethanol, 5-bromo-2-hydroxy,unii-y1uz53815e
75342
4-bromo-2-(hydroxymethyl)phenol
C1=CC(=C(C=C1Br)CO)O
Color White to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 106.0°C to 111.0°C
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(OH)CH2OH
Beilstein 06, 893
Merck Index 12, 1460
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,benzene,chloroform
Formula Weight 203.04
Physical Form Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-hydroxybenzyl alcohol, 98%
Show More Show Less

EINECSNumber : 219-026-0