Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-iodopyrimidine, 97%

Catalog No. AC434250010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC434250010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434250010 Supplier Thermo Scientific Chemicals Supplier No. 434250010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 183438-24-6 | C4H2BrIN2 | 284.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183438-24-6
Molecular Formula C4H2BrIN2
Molecular Weight (g/mol) 284.88
MDL Number MFCD01318111
InChI Key ZEZKXPQIDURFKA-UHFFFAOYSA-N
Synonym 5-bromo-2-iodo-pyrimidine, 2-iodo-5-bromopyrimidine, 5-bromo-2-iodo pyrimidine, pyrimidine, 5-bromo-2-iodo, 5-bromo-2-iodpyrimidine, 5-bromo-2-iodo-1,3-diazine, pubchem6900, 5-brmo-2-iodopyrimidine, acmc-1c1ed, 2-iodo-5-bromo-pyrimidine
PubChem CID 7006651
IUPAC Name 5-bromo-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 100°C to 102°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 284.88
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-iodopyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.