Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B336925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B336925G 25 g
B33695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B336925G Supplier TCI America Supplier No. B336925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80866-82-6
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.062
MDL Number MFCD00004630
InChI Key HAOOGRLZVQWDBY-UHFFFAOYSA-N
PubChem CID 612298
IUPAC Name (5-bromo-2-methoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)Br)CO

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 25 g
Synonym 5-bromo-2-methoxybenzyl alcohol, 5-bromo-2-methoxyphenyl methanol, 5-bromo-2-methoxybenzylalcohol, benzenemethanol, 5-bromo-2-methoxy, 4-bromo-2-hydroxymethyl anisole, 5-bromo-2-methoxy-phenyl-methanol, 2-methoxy-5-bromobenzyl alcohol, 5-bromo-o-anisyl alcohol, 4-bromo-hydroxymethyl anisole, benzenemethanol,5-bromo-2-methoxy
Formula Weight 217.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.