Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B48081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48081G 1 g
B48085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48081G Supplier TCI America Supplier No. B48081G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156001-51-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD08061920
InChI Key WNVUTFDOGUGEIS-UHFFFAOYSA-N
Synonym 4-Bromo-2-cyanotoluene
PubChem CID 12994004
IUPAC Name 5-bromo-2-methylbenzonitrile
SMILES CC1=C(C=C(C=C1)Br)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 196.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.